Titriplex® Lösung A zur Erdalkalibestimmung in Wasser, Titrisol®, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
1.08419.1000
1.08419.1000EA
79.7
EUR
1.08419.1000
Titriplex® Lösung A zur Erdalkalibestimmung in Wasser, Titrisol®, Supelco®
Titriplex® Lösung A
for the determination of alkaline earth metals in water 1 ml ≙ 56 mg CaO/l using 100 ml of water
1 ml of reagent is equivalent to 5,6 degree German hardness when using 100 ml of water.
1°d ≙ 10 mg CaO/l
Spezifikation Testergebnisse
Efficiency value | 5.57 - 5.63 °dH |
Determination method | Complexometric titration |
The efficiency value of this solution was determined with volumetric standard calcium carbonate (article number 1.02410). The expanded measurement uncertainty is ± 0.7 % (k=2 coverage factor for 95% coverage probability). | |
The certified value is traceable to primary standard NIST SRM 682 (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard calcium carbonate, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025. |