Karbolfuchsin -Lösung, AFB-color Färbemittel für Bakteriologie, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidin Hydrochlorid, 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidiniumchlorid, 4-(Di(4-amino-3-methylphenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchlorid, 3,3'-Dimethyl-4,4',4''-triaminotrityliumchlorid

M-Clarity™ Qualitätsstufe = MQ300

1.08512.0500 1.08512.2500
1.08512.0500EA 240 EUR
1.08512.0500 1.08512.2500
Karbolfuchsin -Lösung, AFB-color Färbemittel für Bakteriologie, Sigma-Aldrich®
Karbolfuchsin

Tb-color carbol fuchsin solution is used to detect mycobacteria (AFB cold staining)


Tb-color carbol fuchsin solution is used for microscopic investigation of mycobacteria. This Tb-color staining kit is based on the classic Ziehl-Neelsen staining method. The Tb-color carbol fuchsin solution stains mycobacteria red in microscopic preparations, whereas other microorganisms take on the blue from the blue counter staining. The solution is an IVD product and CE registered.

Formel: C₂₁H₂₂ClN₃
Molecular Weight: 351.88 g/mol
Dichte: 0.99 g/cm3 (20 °C)
Flammpunkt: 48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013129
CAS-Nummer: 4197-24-4
UN: 1992
ADR: 3 (6.1) III
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR