Zinkoxid, EMPROVE® EXPERT BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®

Lieferant: Merck
Achtung

Synonyme: Zink(II)oxid

1.08846.9029 1.08846.1000
1.08846.9029EA 0 EUR
1.08846.9029 1.08846.1000
Zinkoxid, EMPROVE® EXPERT BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Zinkoxid
Formel: ZnO
Molecular Weight: 81.39 g/mol
Dichte: 5.61 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR