Zinksulfat 0.1 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph. Eur. volumetrische Lösung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

1.08879.1000
1.08879.1000EA 39.13 EUR
1.08879.1000
Zinksulfat 0.1 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph. Eur. volumetrische Lösung, Supelco®
Zinksulfat

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: O₄SZn
Molecular Weight: 161,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011302
CAS-Nummer: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR