Schwefelsäure 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.09074.1000 1.09074.5000 1.09074.9010 1.09074.4000 1.09074.9013
1.09074.1000EA 24.68 EUR
1.09074.1000 1.09074.5000 1.09074.9010 1.09074.4000 1.09074.9013
Schwefelsäure 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®
Schwefelsäure

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.

Formel: H₂SO₄
Molecular Weight: 98.08 g/mol
Siedepunkt: 330
Schmelzpunkt: 10,38
Dichte: 1.01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR