Iod 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph. Eur. volumetrische Lösung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

1.09099.1000 1.09099.1003
1.09099.1000EA 44.66 EUR
1.09099.1000 1.09099.1003
Iod 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. Ph. Eur. volumetrische Lösung, Supelco®
Iod

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: I₂
Molecular Weight: 253.81 g/mol
Siedepunkt: 184,3
Schmelzpunkt: 114
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
EINECS: 231-442-4
UN: 3264
Merck Index: 13,05036

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR