Kaliumdichromat 1/24 mol/l (0.25 N) in wässriger Lösung, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.09118.1000
1.09118.1000EA 47.6 EUR
1.09118.1000
Kaliumdichromat 1/24 mol/l (0.25 N) in wässriger Lösung, Supelco®
Kaliumdichromat

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: K₂Cr₂O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00011367
CAS-Nummer: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 3.8 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR