Natriumhydroxid 2 mol/l (2 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.09136.1000 1.09136.9025 1.09136.1003
1.09136.1000EA 22 EUR
1.09136.1000 1.09136.9025 1.09136.1003
Natriumhydroxid 2 mol/l (2 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®
Sodium hydroxide

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: NaOH
Siedepunkt: 109 °C (1013 hPa)
Dichte: ≤1,11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Form liquid
Amount-of-substance concentration 1.990 - 2.010
Measurement uncertainty +/- 0.006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR