Natriumhydroxid 0,2 mol/l (0,2 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

1.09140.1000 1.09140.9010 1.09140.1003
1.09140.1000EA 25.2 EUR
1.09140.1000 1.09140.9010 1.09140.1003
Natriumhydroxid 0,2 mol/l (0,2 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®
Sodium hydroxide

Merck have introduced into their Titripur® Titrants range packs with additional technology - Supelco® SmartTitrants use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. Mettler Toledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (ProductPage) in the Related Information tab below.

Formel: NaOH
Siedepunkt: ∼105 °C (1013 hPa)
Dichte: 1,02 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Form liquid
Amount-of-substance concentration 0.1990 - 0.2010
Measurement uncertainty 0.0006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR