Natriumhydroxid 0.02 mol/l (0.02 N) aqueous solution, Titripur® Volumetric, Supelco®

Lieferant: Merck

Synonyme: Caustic soda

MQ100

1.09142.0500
1.09142.0500EA 27.6 EUR
1.09142.0500
Natriumhydroxid 0.02 mol/l (0.02 N) aqueous solution, Titripur® Volumetric, Supelco®
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
Dichte: 1 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Form liquid
Amount-of-substance concentration 0.01990 - 0.02002
Measurement uncertainty + / - 0.00006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR