Quecksilber(II)nitrat 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. USP, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Quecksilberdinitrat

M-Clarity™ Qualitätsstufe = MQ100

1.09143.1000
1.09143.1000EA 113 EUR
1.09143.1000
Quecksilber(II)nitrat 0.05 mol/l (0.1 N) in wässriger Lösung, Titripur® Reag. USP, Supelco®
Quecksilber(II)nitrat

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: HgN₂O₆
Molecular Weight: 324,60 g/mol
Dichte: 1,025 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011038
CAS-Nummer: 10045-94-0
EINECS: 233-152-3
UN: 3289
ADR: 6.1 (8),II

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Density 1.02 g/cm3 (20 °C)
pH 1 (H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR