Fluorescent indicator F₂₅₄, Millipore®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

1.09182.0050
1.09182.0050EA 78.6 EUR
1.09182.0050
Fluorescent indicator F₂₅₄, Millipore®
Fluorescent indicator F₂₅₄

Application: Fluorescent indicator F₂₅₄. CAS 68611-47-2, pH 7.0 (H₂O) suspension.
Physicochemical Information
Density: 4.0 g/cm3 (20 °C)
Melting Point: 1500 °C
pH value: 7.0 (H₂O) suspension
Bulk density: 1450 kg/m3
Solubility: <0.1 g/l

Lagertemperatur: Raumtemperatur
CAS-Nummer: 68611-47-2
EINECS: 271-896-0

Bestellen

Spezifikation Testergebnisse

Identity Conforms
Fe (iron) ≤0,01%
Water ≤0,3%
Application test for DC Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR