Pufferlösungen, Certipur®, farbcodiert, Supelco®
Lieferant: Merck
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
1.09475.0500EA 44.1 EUR
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
Pufferlösungen, Certipur®, farbcodiert, Supelco®
Puffer pH-Puffer
Lösungen zur genauen Kalibrierung und Überwachung von pH-Messinstrumenten.
  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).