1,4-Dioxan Stabilised, EMSURE® ACS, ISO analysis_for, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 1,4-Diethylendioxid, p-Dioxan, Glycolethylether, 1,4-Dioxacyclohexan

Stabilisierung: 25 ppm BHT
MQ300

1.09671.0250 1.09671.1000 1.09671.2500 1.09671.6025
1.09671.0250EA 56.1 EUR
1.09671.0250 1.09671.1000 1.09671.2500 1.09671.6025
1,4-Dioxan Stabilised, EMSURE® ACS, ISO analysis_for, Supelco®
1,4-Dioxan

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: C₄H₈O₂
Molecular Weight: 88.11 g/mol
Siedepunkt: 101.1
Schmelzpunkt: 11.8
Dichte: 1.03 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006571
CAS-Nummer: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 12 °C
Molar Mass 88.11 g/mol
Boiling Point 101.5 °C (1013 hPa)
Vapor Pressure 41 hPa (20 °C)
Flash Point 11 °C
Explosion Limit 1.7 - 25.2 %(V)
Density 1.03 g/cm3 (20 °C)
pH 6 - 8 (500 g/l, H2O, 20 °C)
Ignition Point 300 °C
Evaporation number 7.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR