Sulphate standard solution, concentrate, 1000 mg SO₄, Titrisol®, Supelco®
Lieferant: Merck
1.09872.0001
1.09872.0001EA
48.2
EUR
1.09872.0001
Sulphate standard solution, concentrate, 1000 mg SO₄, Titrisol®, Supelco®
Standards
Anionische Standards
Sulfat-Standards
Titrisol® standards are traceable to standard reference material from NIST.
Standards: A batch-specific Certificate of Analysis is available.