Ammoniumthiocyanat 0.1 mol/l aqueous solution_concentrated, Titrisol®, Supelco®
Lieferant: Merck
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
MQ100
1.09900.0001
1.09900.0001EA
38.1
EUR
1.09900.0001
Ammoniumthiocyanat 0.1 mol/l aqueous solution_concentrated, Titrisol®, Supelco®
Ammoniumthiocyanat
Formel:
NH₄SCN Molecular Weight: 76.12 g/mol Schmelzpunkt: 150 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Density | 1.03 g/cm3 (20 °C) |
pH | 4.5 (H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...