Iodid-Iodat concentrate for dilution to c(I₂) = 1/128 mol/l (1/64 N) aqueous solution, Titrisol® Volumetric, Supelco®

Lieferant: Merck
Gefahr


MQ100

1.09914.0001
1.09914.0001EA 41.4 EUR
1.09914.0001
Iodid-Iodat concentrate for dilution to c(I₂) = 1/128 mol/l (1/64 N) aqueous solution, Titrisol® Volumetric, Supelco®
Iodid-Iodat

Dose for preparation of 1 litre of solution.

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Density 1.07 g/cm3 (20 °C)
pH 13 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR