Salzsäure 6 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®

Lieferant: Merck
Achtung

Synonyme: Hydrochloric acid solution, Muriatic acid

1.10164.9950 1.10164.1000 1.10164.9025 1.10164.2500 1.10164.9208
1.10164.9950EA 0 EUR
1.10164.9950 1.10164.1000 1.10164.9025 1.10164.2500 1.10164.9208
Salzsäure 6 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Salzsäure

Manufactured from hydrochloric acid fuming 37% EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF and purified water acc. Ph. Eur., JP.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formel: HCl
Molecular Weight: 36.46 g/mol
Dichte: 1.1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8

Bestellen

Spezifikation Testergebnisse

Concentration (HCl) 5.7 - 6.3 mol/l
Identity Passes Test
Color ≤10 Hazen
Appearance Passes Test
Free Bromine or Free Chlorine Passes Test
Free Chlorine (Cl) ≤0.0001
Sulfate (SO₄) ≤0.0005%
Sulfite (SO₃) ≤0.001%
Heavy Metals (as Pb) ≤0.0001%
As (Arsenic) ≤0.00015%
Residual Solvents (ICH Q3C) Excluded by Production Process
Residue on Ignition (as SO₄) ≤0.008%
Evaporation Residue ≤0.01
Endotoxins ≤0.25 I.U./ml
Filtered with 0.2 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR