EC (Escherichia coli) Broth acc. ISO 7251, FDA‑BAM and SMWW GranuCult® plus, Millipore
Lieferant: Merck
1.10765.0500
1.10765.0500EA 262 EUR
1.10765.0500
EC (Escherichia coli) Broth acc. ISO 7251, FDA‑BAM and SMWW GranuCult® plus, Millipore
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
EC (Escherichia coli) broth
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
Media is granulated.