Titan(III)chlorid 15% in Salzsäure 10% Reag. Ph. Eur., Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Titantrichlorid
M-Clarity™ Qualitätsstufe = MQ200
1.10789.0001
1.10789.0001EA
121
EUR
1.10789.0001
Titan(III)chlorid 15% in Salzsäure 10% Reag. Ph. Eur., Sigma-Aldrich®
Titanium(III) chloride
Formel:
Cl₃Ti Molecular Weight: 154,24 g/mol |
MDL:
MFCD00011266 CAS-Nummer: 7705-07-9 EINECS: 231-728-9 UN: 1789 ADR: 8, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...