Ethylacetat, SupraSolv® GC-ECD/FID, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Essigester, Essigsäureethylester, EtOAc

MQ100

1.10972.1000 1.10972.2500 1.10972.4000 1.10972.9030
1.10972.1000EA 47.83 EUR
1.10972.1000 1.10972.2500 1.10972.4000 1.10972.9030
Ethylacetat, SupraSolv® GC-ECD/FID, Supelco®
Ethylacetat

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionisation detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formel: CH₃COOC₂H₅
Molecular Weight: 88.11 g/mol
Siedepunkt: 77.1
Schmelzpunkt: –83 °C
Dichte: 0.902 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 85.3 g/l (20 °C)
Melting Point -83 °C
Molar Mass 88.11 g/mol
Boiling Point 77 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point -4 °C
Refractive Index 1.37
Explosion Limit 2.1 - 11.5 %(V)
Density 0.90 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 460 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR