3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
M-Clarity™ Qualitätsstufe = MQ100
1.11714.0001
1.11714.0010
1.11714.0001EA
236
EUR
1.11714.0001
1.11714.0010
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid, Sigma-Aldrich®
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formel:
C₁₈H₁₆N₅BrS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011964 CAS-Nummer: 298-93-1 EINECS: 206-069-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...