Ethanol ≥99,9% (durch GC), LiChrosolv®, Gradienten-Qualität für die Flüssigchromatographie, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.
Formel:
C₂H₅OH Siedepunkt: 78,3 °C (1013 hPa) Schmelzpunkt: –117 °C Dichte: 0,7895 g/cm³ (20 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3, REACH: 01-2119457610-43 Merck Index: 12,03806 |
Spezifikation Testergebnisse
Purity (GC) | ≥ 99.9 % |
Identity (IR) | conforms |
Evaporation residue | ≤ 2.0 mg/l |
Water | ≤ 0.1 % |
Acidity | ≤ 0.0002 meq/g |
Alkalinity | ≤ 0.0002 meq/g |
Gradient grade (at 235 nm) | ≤ 5 mAU |
Gradient grade (at 254 nm) | ≤ 2 mAU |
Transmission (at 225 nm) | ≥ 60 % |
Transmission (at 240 nm) | ≥ 85 % |
Transmission (from 260 nm) | ≥ 98 % |
Filtered by 0.2 µm filter | |
Synonyms | Ethyl alcohol, EtOH |
Hill Formula | C2H6O |
Chemical Formula | C2H5OH |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...