Gram-color Färbekit, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.11885.0001
1.11885.0001EA 218 EUR
1.11885.0001
Gram-color Färbekit, Sigma-Aldrich®
Gram color staining kit

Gram-color Färbekit, beinhaltet je 500 ml Kristallviolet Lösung, Lugol's Lösung stabilisiert, Safranin Lösung und 2× 500 ml Entfärber


In bacteriology, Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The murein structure of the bacteria wall is the basis of the colour affinity. In the first step, bacteria is stained with crystal violet, an aniline dye. After treatment with iodine solution (Lugol’s solution), a dye-iodine complex is formed. During the decolourising step, this complex stays in the multi-layer murein structure of the cell wall of Gram-positive bacteria - they appear blue. Gram-negative bacteria have a monolayer murein structure, the dye-iodine complex does not stay bound to the cell wall - they are decolourised. Gram-negative bacteria can then be counterstained with safranin solution and this will then appear orange. The Gram-colour stain set contains 5×500 ml safe PE bottles and is sufficient for staining up to 250 slides. It fulfils all quality requirements for IVD registration and CE certification.

Lagertemperatur: Raumtemperatur
UN: 1993
ADR: 3,
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR