Hemacolor® Schnelles Färben von Blutausstrich -Lösung I fixatives Reagens, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Fixing solution (contains methanol)
Hemacolor® can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, sperm, effusions and lavages, and in addition, for blood and bone marrow smears. The Hemacolor® staining kit is available with 100 ml or 500 ml bottles. It is developed and registered as CE / IVD product for applications in diagnostics. All Hemacolor solutions are additionally available as single 2.5 l PE-bottles. The package is sufficient for 1250 - 5000 applications. The buffer tablets acc. to Weise pH 7.2 are also available in larger pack sizes, thus, they can be useful for higher demands:
Hemacolor® solution 1, fixing solution
Hemacolor® solution 2, color reagent red
Hemacolor® solution 3, color reagent blue
Lagertemperatur:
Raumtemperatur |
UN:
1230 ADR: 3 (6.1), |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...