Tellurdioxid ≥99,5%, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

1.12356.1000
1.12356.1000EA 2720 EUR
1.12356.1000
Tellurdioxid ≥99,5%, Sigma-Aldrich®
Tellurdioxid
Formel: O₂Te
MDL: MFCD00011263
CAS-Nummer: 7446-07-3

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 733 °C
Molar Mass 159.60 g/mol
Boiling Point 1245 °C
Density 6.02 g/cm3 (20 °C)
Synonyms Tellurium(IV) oxide
Hill Formula O2Te
Chemical Formula TeO2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR