Acetonitril für die DNA-Synthese

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.13212.2500 1.13212.4000 1.13212.9185
1.13212.2500EA 219 EUR
1.13212.2500 1.13212.4000 1.13212.9185
Acetonitril für die DNA-Synthese
Acetonitril

Additional to our high quality acetonitrile dried with low water content up to 10 ppm our DNA-/RNA synthesis reagents with its high quality, lowest water and acid content are best requirements for a reliable and reproducible DNA-/RNA synthesis with high yields. A wide variety of packaging types supporting specific synthesis instrumentations, for quantities from bottle to bulk, and the will to customized solutions completes our innovative offering.

Formel: H₃CCN
Siedepunkt: 81,6 °C (1013 hPa)
Schmelzpunkt: –45,7 °C
Dichte: 0,782 g/cm³ (20 °C)
Flammpunkt: 2 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point -45.7 °C
Molar Mass 41.05 g/mol
Boiling Point 81.6 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point 2 °C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 524 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR