Acetonitril, Prepsolv® für die präparative Chromatographie
Lieferant: Merck
|
Gefahr
|
Synonyme:
Ethannitril, Essigsäurenitril, Ethansäurenitril, Methylcyanid
M-Clarity™ Qualitätsstufe = MQ200
1.13358.2500
1.13358.9030
1.13358.9185
1.13358.2509
1.13358.2500EA
285
EUR
1.13358.2500
1.13358.9030
1.13358.9185
1.13358.2509
Acetonitril, Prepsolv® für die präparative Chromatographie
Acetonitril
Prepsolv® solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.
Formel:
H₃CCN Molecular Weight: 41.05 g/mol Siedepunkt: 81.6 °C (1013 hPa) Schmelzpunkt: –45.7 °C Dichte: 0.782 g/cm³ (20 °C) Flammpunkt: 2 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...