Lactophenol blue solution Farbstoff (pilzartig), Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.13741.0100
1.13741.0100EA 29.2 EUR
1.13741.0100
Lactophenol blue solution Farbstoff (pilzartig), Sigma-Aldrich®
Lactophenol blue solution

Lactophenol blue solution for staining fungi.


Lactophenol blue solution is ready-to-use staining solution for fungi staining in bacteriology with samples of human origin. The material is stained in one step with Lactophenol blue solution. Fungi appear dark blue. This ready-to-use solution is especially for air-dried, heat-fixed smears of bacteriological material, e.g. sputum, FNAB, lavages, imprints, body fluids, puss, and exudates, liquid and solid cultures.
The staining solution is sufficient for up to 50 preparations per bottle. It is developed and registered as CE / IVD product for applications in diagnostics.

Dichte: 1,16 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00146574
UN: 2821
ADR: 6.1,
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR