Dihexyl sodium sulphosuccinate zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

1.13861.0025
1.13861.0025EA 671 EUR
1.13861.0025
Dihexyl sodium sulphosuccinate zur Synthese, Sigma-Aldrich®
Dihexyl sodium sulphosuccinate
Formel: C₁₆H₂₉NaO₇S
Molecular Weight: 388,451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050501
CAS-Nummer: 3006-15-3
EINECS: 221-109-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e87675f7537dae777b34f07f3406f80065dc1f53&vid=e87675f7537dae777b34f07f3406f80065dc1f53&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 364 g/l
Melting Point 230 °C decomposes
Molar Mass 388.46 g/mol
Bulk Density 180 kg/m3
pH 5 - 7 (50 g/l, H2O, 25 °C)
Bulk density 180 kg/m3
Storage +15...+25 °C
Assay ≥98,5%
Identity Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR