Standard oil für die Atomabsorptionsspektroskopie, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
1.13898.2500
1.13898.2500EA
539
EUR
1.13898.2500
Standard oil für die Atomabsorptionsspektroskopie, Supelco®
Standard oil
Standard oil, solvent for atomic absorption spectroscopy
Dichte:
0,85 g/cm3 (20 °C) Flammpunkt: >180 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
8042-47-5 |
Spezifikation Testergebnisse
Identity (IR-spectrum) | passes test |
Density (d 20 °C/ 4 °C) | 0.84 - 0.86 g/ml |
Viscosity (20 °C) | 30 - 40 mPa∙s |
Ag (Silver) | ≤ 0.5 ppm |
Al (Aluminium) | ≤ 0.5 ppm |
Au (Gold) | ≤ 0.5 ppm |
B (Boron) | ≤ 0.5 ppm |
Ba (Barium) | ≤ 0.5 ppm |
Be (Beryllium) | ≤ 0.5 ppm |
Bi (Bismuth) | ≤ 0.5 ppm |
Ca (Calcium) | ≤ 0.5 ppm |
Cd (Cadmium) | ≤ 0.5 ppm |
Co (Cobalt) | ≤ 0.5 ppm |
Cr (Chromium) | ≤ 0.5 ppm |
Cu (Copper) | ≤ 0.5 ppm |
Fe (Iron) | ≤ 0.5 ppm |
Ga (Gallium) | ≤ 0.5 ppm |
In (Indium) | ≤ 0.5 ppm |
K (Potassium) | ≤ 0.5 ppm |
Li (Lithium) | ≤ 0.5 ppm |
Mg (Magnesium) | ≤ 0.5 ppm |
Mn (Manganese) | ≤ 0.5 ppm |
Mo (Molybdenum) | ≤ 0.5 ppm |
Na (Sodium) | ≤ 0.5 ppm |
Ni (Nickel) | ≤ 0.5 ppm |
Pb (Lead) | ≤ 0.5 ppm |
Pt (Platinum) | ≤ 0.5 ppm |
Sb (Antimony) | ≤ 0.5 ppm |
Sn (Tin) | ≤ 0.5 ppm |
Sr (Strontium) | ≤ 0.5 ppm |
Ti (Titanium) | ≤ 0.5 ppm |
Tl (Thallium) | ≤ 0.5 ppm |
V (Vanadium) | ≤ 0.5 ppm |
Zn (Zinc) | ≤ 0.5 ppm |
Zr (Zirconium) | ≤ 0.5 ppm |
Water | ≤ 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...