Acetonitril, LiChrosolv®, isokratische Qualität für die Flüssigchromatographie, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
1.14291.1000EA 146 EUR
1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
Acetonitril, LiChrosolv®, isokratische Qualität für die Flüssigchromatographie, Supelco®
Acetonitril

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formel: H₃CCN
Siedepunkt: 81,6 °C (1013 hPa)
Dichte: 0,782 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR