HPTLC plates silica gel 60, RP-modified layers, glass backed
Lieferant: Merck
1.05914.0001 1.13124.0001 1.13724.0001 1.13725.0001 1.13726.0001 1.14296.0001 1.16225.0001
1.05914.0001EA 861 EUR
1.05914.0001 1.13124.0001 1.13724.0001 1.13725.0001 1.13726.0001 1.14296.0001 1.16225.0001
HPTLC plates silica gel 60, RP-modified layers, glass backed
TLC-Platten
Our NH₂, CN- and DIOL-modified silica sorbents are less polar then conventional silica phases, making them ideal for separating hydrophilic or charged substances. Our moderately polar cyano- and diol-modified silica plates can be used for both normal phase and reversed phase systems. An alternative to PEI cellulose, amino-modified NH₂ plates provide weak basic ion exchange characteristics with special selectivity for charged compounds.
HPTLC concentrating zone plates allow easy application of large volumes of diluted samples. They include a purification and concentration step.