Schnelltest-Kits, Chromat, MQuant®, Supelco®
Lieferant: Merck
1.14402.0001 1.14441.0001 1.14756.0001
1.14402.0001EA 156 EUR
1.14402.0001 1.14441.0001 1.14756.0001
Schnelltest-Kits, Chromat, MQuant®, Supelco®
Umwelt-Prüfkits
Schnelltests, Visueller Farbvergleich.
  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® mit Farbkartenkomparator (MCC)
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
MQuant® mit Farbscheibenkomparator (MCD)
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
Lieferumfang: Jedes Kit beinhaltet die notwendigen Reagenzien, Küvetten, Farbvergleichsübersicht oder Drehscheiben-Komparator, Bedienungsanleitung.