Wasser, LiChrosolv® für die Chromatographie, für die LC-MS, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.15333.1000 1.15333.2500 1.15333.9010 1.15333.9030 1.15333.9185 1.15333.4000
1.15333.1000EA 15.5 EUR
1.15333.1000 1.15333.2500 1.15333.9010 1.15333.9030 1.15333.9185 1.15333.4000
Wasser, LiChrosolv® für die Chromatographie, für die LC-MS, Supelco®
Wasser

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR