2,2,4-Trimethylpentan , SupraSolv® GC-ECD/FID, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Isooktan, Isobutyltrimethylmethan

MQ100

1.15440.1000 1.15440.2500
1.15440.1000EA 119 EUR
1.15440.1000 1.15440.2500
2,2,4-Trimethylpentan , SupraSolv® GC-ECD/FID, Supelco®
Isooctan

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formel: (CH₃)₂CHCH₂C(CH₃)₃
Molecular Weight: 114.23 g/mol
Siedepunkt: 99.2
Schmelzpunkt: –107 °C
Dichte: 0.692 g/cm³ (20 °C)
Flammpunkt: –12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008943
CAS-Nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 0.56 mg/l (25 °C)
Melting Point -107 °C
Molar Mass 114.23 g/mol
Boiling Point 99 °C (1013 hPa)
Vapor Pressure 51 hPa (20 °C)
Flash Point -14 °C
Explosion Limit 1 - 5.5 %(V)
Density 0.69 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 410 °C
Saturation concentration 239 g/m3 (20 °C)
Evaporation number 2.3
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR