Coomassie Brillantblau G 250, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylen]-3-methylcyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau G 250, Brillantindocyanin G, Acid Blue 90, C.I. 42655, Brillantblau G
M-Clarity™ Qualitätsstufe = MQ100
1.15444.0025
1.15444.0025EA
64.8
EUR
1.15444.0025
Coomassie Brillantblau G 250, Sigma-Aldrich®
Coomassie Brillantblau G 250
Formel:
C₄₇H₄₈N₃NaO₇S₂ Molecular Weight: 854.04 g/mol Dichte: Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078482 CAS-Nummer: 6104-58-1 EINECS: 228-058-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...