Hyamine® 1622 0.004 mol/l, Titripur® volumetrische Lösung, zur Bestimmung von anionischen Tensiden, Supelco®

Lieferant: Merck

Synonyme: Hyamine® 1622

M-Clarity™ Qualitätsstufe = MQ100

1.15480.1000 1.15480.1003
1.15480.1000EA 37.5 EUR
1.15480.1000 1.15480.1003
Hyamine® 1622 0.004 mol/l, Titripur® volumetrische Lösung, zur Bestimmung von anionischen Tensiden, Supelco®
Benzethoniumchlorid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: C₂₇H₄₂ClNO₂
Dichte: 1,0 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011742
CAS-Nummer: 121-54-0
EINECS: 204-479-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR