Aldehyde dehydrogenase (from Yeast)

Lieferant: Merck

1.15640.0001
1.15640.0001EA 177 EUR
1.15640.0001
Aldehyde dehydrogenase (from Yeast)
Enzyme

  • From Yeast
  • E.C.1.2.1.5
  • Freeze dried stabilized powder
  • Activity: 2 unit/mg solid (approx. 20 u/mg protein)
  • ADH: ≤0,001 U/mg
  • LDH: ≤0,001 U/mg


Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 9,0 and 25 °C.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR