Hämatoxylin Monohydrat, Certistain® für die Mikroskopie, Sigma-Aldrich®

Lieferant: Merck

1.15938.0025 1.15938.0100 1.15938.1000
1.15938.0025EA 156 EUR
1.15938.0025 1.15938.0100 1.15938.1000
Hämatoxylin Monohydrat, Certistain® für die Mikroskopie, Sigma-Aldrich®
Hämatoxylin Monohydrat

Hematoxylin-Monohydrat (C.I. 75290) Certistain® is a dye for the specific staining of DNA in the nuclei. It is used in cytology and histology. Nuclei are stained blue, dark violet to black. Hematoxylins are mixed with trivalent positively charged metal salts and build up the so-called hematoxylin lakes, used for the selective staining of nuclei (DNA). Certistain® is the quality group for seleceted high quality dyes that fulfill all quality requirements for IVD and CE registration.

Formel: C₁₆H₁₄O₆·H₂O
CAS-Nummer: 517-28-2
EINECS: 208-237-3
IVDLogo

Bestellen

Spezifikation Testergebnisse

Hill Formula C₁₆H₁₄O₆ * H₂O
Molar Mass 320.29 g/mol
Melting Point 140 °C Elimination of water of crystallisation
Bulk density 500 kg/m³
Solubility (20 °C) slightly soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR