Eisenhämatoxylin kit (Weigert's) Stain_histology, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Weigert's solution, Weigert's stain

1.15973.0002
1.15973.0002EA 281 EUR
1.15973.0002
Eisenhämatoxylin kit (Weigert's) Stain_histology, Sigma-Aldrich®
Eisenhämatoxylin

Nuclear staining in connective tissue staining methods


ron hematoxylin solutions are used for staining nuclei of tissue sections, e.g. elastic fibers stained according to van Gieson or connective tissue stained by various trichromo staining techniques, where acidic stains were used as counter-stain. As commonly used hemalum solutions, such as Mayer’s Hemalum, Delafield, Gill or Harris, only weakly stain nuclei when used with acidic counterstains, the Weigert's Iron hematoxylin stain is an effective and robust alternative. The kit contains two solutions; an alcoholic hematoxylin solution and a hydrochloric acid iron(III)nitrate solution, which are ready-to-use and should be applied at 3-5 µm paraffin sections. Weigert's Iron hematoxylin kit is an IVD & CE product and can be used in the clinical laboratory for diagnostics.The package is sufficient for 400 - 500 applications.

Lagertemperatur: Raumtemperatur
MDL: MFCD00284879
UN: 1170
ADR: 3
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR