Eshmuno® CPX, Resin Racemat_separation, Chromatography
Lieferant: Merck
|
Achtung
|
1.20083.0100
1.20083.0500
1.20083.0100_T
1.20083.0500_T
1.20083.5000
1.20083.0100EA
1510
EUR
1.20083.0100
1.20083.0500
1.20083.0100_T
1.20083.0500_T
1.20083.5000
Eshmuno® CPX, Resin Racemat_separation, Chromatography
Eshmuno® CPX
Eshmuno® CPX media is a strong cation exchanger built on the proven Eshmuno® resin technology.
- Excellent mAb monomer/aggregate separation efficiency
- High resolution intermediate purification
- Superior dynamic binding capacity
- High throughput purification
- Rigid base bead for easy packing
- Outstanding pressure flow behavior
Lagertemperatur:
Raumtemperatur |
UN:
1170 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...