Eshmuno® CPX, Resin Racemat_separation, Chromatography

Lieferant: Merck
Achtung

1.20083.0100 1.20083.0500 1.20083.0100_T 1.20083.0500_T 1.20083.5000
1.20083.0100EA 1510 EUR
1.20083.0100 1.20083.0500 1.20083.0100_T 1.20083.0500_T 1.20083.5000
Eshmuno® CPX, Resin Racemat_separation, Chromatography
Eshmuno® CPX

Eshmuno® CPX media is a strong cation exchanger built on the proven Eshmuno® resin technology.


  • Excellent mAb monomer/aggregate separation efficiency
  • High resolution intermediate purification
  • Superior dynamic binding capacity
  • High throughput purification
  • Rigid base bead for easy packing
  • Outstanding pressure flow behavior

Lagertemperatur: Raumtemperatur
UN: 1170

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR