ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2), Millipore®

Lieferant: Merck

Synonyme: Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, NADP-Na2, ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, Nadidphosphat Dinatriumsalz, NADP Dinatriumsalz

M-Clarity™ Qualitätsstufe = MQ100

1.24541.0001
1.24541.0001EA 352 EUR
1.24541.0001
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2), Millipore®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Formel: C₂₁H₂₆N₇Na₂O₁₇P₃
Molecular Weight: 787,37 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065390
CAS-Nummer: 24292-60-2
EINECS: 246-129-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR