Ammonium CRMs, standard solutions, Supelco®
Lieferant: Merck
1.25023.0100
1.25024.0100
1.25022.0100
1.25025.0100
1.25026.0100
1.25027.0100
1.32227.0100
1.25023.0100EA
52.1
EUR
1.25023.0100
1.25024.0100
1.25022.0100
1.25025.0100
1.25026.0100
1.25027.0100
1.32227.0100
Ammonium CRMs, standard solutions, Supelco®
Standards
Kationischer Standards
Ammonium-Standards
NH₄Cl in H₂O
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.