Nitrogen (total) CRM, Standard Solutions, Supelco®
Lieferant: Merck
1.25043.0100 1.25044.0100 1.25045.0100
1.25043.0100EA 52.1 EUR
1.25043.0100 1.25044.0100 1.25045.0100
Nitrogen (total) CRM, Standard Solutions, Supelco®
Standards Umweltanalyse-Standards
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.