Aluminium CRM, Standard Solutions, Supelco®
Lieferant: Merck
1.32225.0100
1.32225.0100EA
52.1
EUR
1.32225.0100
Aluminium CRM, Standard Solutions, Supelco®
Standards
Kationischer Standards
Aluminium-Standards
Al(NO₃)₃ in H₂O
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= ICP-OES
Standards: Traceable to SRM from NIST and USP.