Fluoride CRM, Standard Solutions, Supelco®
Lieferant: Merck
1.32233.0100
1.32234.0100
1.32235.0100
1.32236.0100
1.32233.0100EA
90.7
EUR
1.32233.0100
1.32234.0100
1.32235.0100
1.32236.0100
Fluoride CRM, Standard Solutions, Supelco®
Standards
Anionische Standards
Fluorid-Standards
NaF in H₂O
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.