Nitrate CRM, standard solutions, Supelco®
Lieferant: Merck
1.25036.0100
1.25037.0100
1.25038.0100
1.25039.0100
1.25040.0100
1.32240.0100
1.32241.0100
1.32242.0100
1.25036.0100EA
52.1
EUR
1.25036.0100
1.25037.0100
1.25038.0100
1.25039.0100
1.25040.0100
1.32240.0100
1.32241.0100
1.32242.0100
Nitrate CRM, standard solutions, Supelco®
Standards
Anionische Standards
Nitrat-Standards
NaNO₃ in H₂O
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.