Essigsäure glacial_AcOH 100%, EMPROVE® EXPERT JP, BP BiopharmaceuticalProduction, SAFC®

Lieferant: Merck
Gefahr

Synonyme: Ethansäure

1.37000.2500 1.37000.9025 1.37000.5000 1.37000.9191 1.37000.9950
1.37000.2500EA 370 EUR
1.37000.2500 1.37000.9025 1.37000.5000 1.37000.9191 1.37000.9950
Essigsäure glacial_AcOH 100%, EMPROVE® EXPERT JP, BP BiopharmaceuticalProduction, SAFC®
Essigsäure

Bitte beachten Store at +15°C to +25°C.

Formel: H₃CCOOH
Molecular Weight: 60.05 g/mol
Siedepunkt: 118
Schmelzpunkt: 17
Dichte: 1.05 g/cm³ (20 °C)
Flammpunkt: 38,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3)
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric) 99.8 - 100.5 %
Identity passes test
Colour (Hazen) ≤ 10 Hazen
Appearance of the substance passes test
Formic acid, Formates and other oxidable impurities (as HCOOH) ≤ 0.5 %
Titratable base ≤ 0.0004 meq/g
Solidification temperature ≥ 16.3 °C
Chloride (Cl) ≤ 2 ppm
Sulfate (SO₄) ≤ 5 ppm
Heavy metals (as Pb) ≤ 5 ppm
Fe (Iron) ≤ 0.2 ppm
Other residual solvents (ICH Q3C) ...excluded by production process
Reducing substances (Ph Eur) passes test
Substances reducing potassium permanganate passes test
Evaporation residue ≤ 50 ppm
Dilution test passes test
Endotoxins ≤ 2.5 I.U./ml
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR